Learn More
Socorex™ White Capillary Tubes
Make a connection to laboratory pipettes with Socorex™ White Capillary Tubes. Each tube is supplied in a practical box with a lid.
Brand:  Socorex™ 341.025
 View more versions of this product
Product Code. 10330221
  • 88.90€ / Pack of 200
Estimated Shipment: 15-05-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • White
  • Replacement glass capillaries
  • For Acura™ 846 positive displacement pipettes
  • Color coded to match corresponding pipette
Specifications
Specifications
5 to 25 μL
White
Capillary Tube
Acura 846 Positive Displacement Pipette
Sterile
200/Pk.
Single-use
Glass
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Socorex™ White Capillary Tubes > Capacity: 5 to 25μL