Learn More
Unity Lab Services™ Verification Report Balance
ULS offers a full range of support services and programs for all instruments and supplies, regardless of manufacturer. Maximize your technology investments by anticipating and avoiding problems before they occur.
Brand: Unity Lab Services™ UMV-P9309
Description
- For non-regulated weighing scales with a range between 30g and 200Kg
- Verification according to NF EN 45501
- Fidelity
- Excentration
- Linearity and accuracy
- Verification using weights certified by a COFRAC accredited laboratory
Specifications
Balances | |
Verification Report |