Learn More
Thermo Scientific™ Fluid Transfer Assembly, Y Set
Designed to aid many common shop floor processes such as sampling, transfer, and filling, a fit-for-purpose solution in the required tubing size and length can be easily selected from these designs.
Brand: Thermo Scientific™ SH31109.01
This item is not returnable.
View return policy
Description
Standard Fluid Transfer Assembly products interconnect with Thermo Scientific standard Labtainer BPCs, Productainer BPCs, single-use bioreactors, mixers, and fermentors right on the shop floor to deliver a comprehensive and complete set of single-use products to meet the needs of your facility or process.- One set of stocked products reduces the number of customized stock keeping units (SKUs), simplifies the ordering process, and reduces product storage requirements
- Multiple manufacturing sites ensure security in product supply
- Produced under cGMP conditions in ISO 7 clean areas to ensure a high level of quality
- Characterized using ISO, USP, EP, and ASTM test methods to validate performance
Specifications
3 x 6.35 mm (1/4 in.) ID x 11.11 mm (7/16 in.) wall; TPE 374; 2 x 0.51 m (20 in.) to 0.51 m (20 in.); MPC in.sert-body | |
Y Set |
Product Suggestions
Customers who viewed this item also viewed
Product Content Correction
Product Title
Thermo Scientific™ Fluid Transfer Assembly, Y Set > 3 x 6.35 mm (1/4 in.) ID x 11.11 mm (7/16 in.) wall; TPE 374; 2 x 0.51 m (20 in.) to 0.51 m (20 in.); MPC in.sert-body