Learn More
BD Vacutainer™ Venous Blood Collection Tubes: SST™ Serum Separation Tubes
Ideal for use in serum determinations in chemistry
479.00€
Specifications
Material | Plastic |
---|---|
For Use With (Application) | For use in serum determinations in chemistry |
Product Code | Brand | Capacity (Metric) | Closure Color | Additive | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Closure Color | Additive | Price | Quantity & Availability | |||
15866161
|
BD
367985 |
10 mL | Red/Gray | Polymer Gel/Silica Activator |
479.00€
Pack of 1000 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
Description
- Contain spray-coated silica and a polymer gel for serum separation
- Provide an efficient means of serum sample preparation and help to improve laboratory workflow
- Tubes marked as Best Practice Formulary meet demanding requirements of complex diagnostic instruments and current clinical guidelines
- Available with gold conventional tube stopper or red/gray Hemogard™ tube closure that prevents human contact with blood on the stopper or tube rim
Evacuated tubes simplify blood collection
- Blood is drawn directly from the vein into the evacuated sterile collection tubes
- Only one venipuncture is needed to draw multiple samples
- Tubes can be changed while needle and plastic holder remain in place on patient's arm
- Closures are color coded
- Made of polyethylene terephthalate (PET)
- Provide a safer work environment for all personnel by (1) reducing risk of tube breakage during handling; (2) eliminating breakage when tubes are centrifuged at higher speeds; and (3) eliminating breakage or chipping of tube rims when stopper is removed and reinserted
- In addition to reducing potential injuries, fewer tube breakages result in less workflow interruption and fewer patient redraws, providing smoother day-to-day operation
Hemogard™ Stoppers
- Safety shield designed for safer tube handling
- Rubber stopper is recessed inside the plastic shield, so any drops left by a blood collection needle remain isolated from potential contact
- Shield is designed to protect lab personnel from direct contact with blood on the stopper or around the outer rim of the tube, as well as from blood splattering upon opening the tube
Specifications
Plastic | |
For use in serum determinations in chemistry |
Product Title
BD Vacutainer™ Venous Blood Collection Tubes: SST™ Serum Separation Tubes