missing translation for 'onlineSavingsMsg'
Learn More

BD Vacutainer™ SST™ II Advance Tubes

Prevents the contamination of the serum from the separated cellular components. BD Vacutainer™ SST™ II Advance Tubes feature an inert, stable gel that separates the serum and the blood clot during centrifugation. The type of gel used enables short centrifugation times.

Brand:  BD 367953


 View more versions of this product

Product Code. 10829270

  • 95.70€ / Pack of 100

Please to purchase this item. Need a web account? Register with us today!

Edge
Explore more special offers
This item is not returnable. View return policy

Description

Description

Using BD SST™ II Advance, routine analytes in clinical chemistry, such as potassium and glucose, are still stable after a week of storage at 2–8°C. Clinical evaluation of special chemistries (such as therapeutic drugs, proteins, peptides, steroids and vitamins) demonstrates a high degree of stability with the acrylic gel in these tubes, which is a distinct advantage during transport and storage.

Advantages of gel tubes versus non-gel tubes:

  • Stable barrier between serum and clotted blood, therefore better analyte stability and sample quality
  • Optimization of the work flow: short centrifugation time, sample processing and archiving in the primary tube
  • No possibility of misidentification due to the use of secondary tubes

Tube usage guidelines:

  • Clotting Times: the minimum recommended coagulation time for patients who have not received anti-coagulation treatment is 30 minutes. Tubes contain a clot activator of silica particles.
  • Centrifugation Conditions: 1300–2000 g for 10 minutes or alternately 3000 g for 5 minutes, at 18–25°C.
  • Effects of Temperature: cooling of the tube during centrifuging can affect the movement capability of the gel. Optimum separation of serum and coagulated blood is achieved at 20–25°C.
  • Store at 4–25°C and protected from direct sunlight during storage.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Serum Separator Tube
PET
Hemogard
Gold
Used to obtain and separate a serum sample
16 mm
Silica
Sterile
8.5 mL
100 mm
Paper
CE
Product Suggestions

Product Suggestions

Videos
Safety and Handling
Show More
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
BD Vacutainer™ SST™ II Advance Tubes > Capacity: 8.5mL

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.