Learn More
BD Vacutainer™ K2EDTA Tubes
Brand:  BD 368841
 View more versions of this product
Product Code. 15744718
  • 195.00€ / Pack of 1000
Explore more special offers
Specifications
K2 EDTA Tube
PET
2 mL
13 x 75 mm
Paper
K2 EDTA
Hemogard
Pink
Used to obtain either a serum sample or a whole blood or EDTA plasma sample
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Show More
SDS
Documents
Documents
Certificates
Promotions
Promotions
Product Content Correction
Product Title
BD Vacutainer™ K2EDTA Tubes > Capacity: 2mL