Learn More
Tris Hydrochloride, Molecular Biology Grade, Ultrapure, Thermo Scientific™
67.83€ - 1900.00€
Chemical Identifiers
CAS | 1185-53-1 |
---|---|
Molecular Formula | C4H12ClNO3 |
Molecular Weight (g/mol) | 157.594 |
MDL Number | MFCD00012590 |
InChI Key | QKNYBSVHEMOAJP-UHFFFAOYSA-N |
Synonym | Tris(hydroxymethyl)amino methane hydrochloride |
PubChem CID | 93573 |
IUPAC Name | 2-amino-2-(hydroxymethyl)propane-1,3-diol;hydrochloride |
SMILES | C(C(CO)(CO)N)O.Cl |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||||
15855198
|
Thermo Scientific
J22676.36 |
500 g | Plastic bottle | N/A | N/A | N/A | |||||
15885348
|
Thermo Scientific
J22676.A1 |
1 kg | Plastic bottle | N/A | N/A | N/A | |||||
15895348
|
Thermo Scientific
J22676.A7 |
5 kg | Plastic bottle | N/A | N/A | N/A | |||||
15805358
|
Thermo Scientific
J22676.C4 |
25 kg | Plastic bottle | N/A | N/A | N/A | |||||
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
1185-53-1 | |
157.594 | |
QKNYBSVHEMOAJP-UHFFFAOYSA-N | |
93573 | |
C(C(CO)(CO)N)O.Cl |
C4H12ClNO3 | |
MFCD00012590 | |
Tris(hydroxymethyl)amino methane hydrochloride | |
2-amino-2-(hydroxymethyl)propane-1,3-diol;hydrochloride |
Specifications
1185-53-1 | |
Crystalline Powder | |
MFCD00012590 | |
Tris(hydroxymethyl)amino methane hydrochloride | |
C(C(CO)(CO)N)O.Cl | |
157.594 | |
157.6 | |
Tris hydrochloride |
∼152°C (decomposition) | |
C4H12ClNO3 | |
3675235 | |
QKNYBSVHEMOAJP-UHFFFAOYSA-N | |
2-amino-2-(hydroxymethyl)propane-1,3-diol;hydrochloride | |
93573 | |
Molecular Biology |
Safety and Handling
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
missing translation for 'einecsNumber' : 214-684-5
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures