missing translation for 'onlineSavingsMsg'
Learn More
Tris Hydrochloride, Molecular Biology Grade, Ultrapure, Thermo Scientific™ Product Code.: 15895348

Tris Hydrochloride, Molecular Biology Grade, Ultrapure, Thermo Scientific™

Product Code. 15895348 Shop All Thermo Scientific Products
5 kg, Plastic bottle
Click to view available options
Quantity:
500 g
1 kg
5 kg
25 kg
Packaging:
Plastic bottle
This item is not returnable. View return policy

Product Code. 15895348

Brand: Thermo Scientific J22676.A7

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1185-53-1
Molecular Formula C4H12ClNO3
Molecular Weight (g/mol) 157.594
MDL Number MFCD00012590
InChI Key QKNYBSVHEMOAJP-UHFFFAOYSA-N
Synonym Tris(hydroxymethyl)amino methane hydrochloride
PubChem CID 93573
IUPAC Name 2-amino-2-(hydroxymethyl)propane-1,3-diol;hydrochloride
SMILES C(C(CO)(CO)N)O.Cl

Specifications

Melting Point ∼152°C (decomposition)
Physical Form Crystalline Powder
Quantity 5 kg
Packaging Plastic bottle
Beilstein 3675235
Formula Weight 157.6
Grade Molecular Biology
Chemical Name or Material Tris hydrochloride
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Tris Hydrochloride, Molecular Biology Grade, Ultrapure, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.