Learn More
TMIGD2 Antibody (953743), Janelia Fluor™ 646, Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals FAB83162J
Product Code. 18523529
  • 670.00€ / 0.10mL
Explore more special offers
Description
Description
TMIGD2 Monoclonal antibody specifically detects TMIGD2 in Human samples. It is validated for Flow Cytometry, Blockade of Receptor-ligand Interaction, CyTOF
TRUSTED_SUSTAINABILITY
Specifications
Specifications
TMIGD2
Monoclonal
Janelia Fluor 646
50mM Sodium Borate
Mouse
Protein A or G purified from hybridoma culture supernatant
RUO
126259
Store at 4°C in the dark.
IgG2a
Flow Cytometry, Blockade of Receptor-ligand Interaction, CyTOF
953743
Flow Cytometry, Blockade of Receptor-ligand Interaction, CyTOF-ready
CD28H, IGPR1, IGPR-1, MGC23244, transmembrane and immunoglobulin domain containing 2, transmembrane and immunoglobulin domain-containing protein 2
Human embryonic kidney cell line HEK-293-derived human TMIGD2/CD28H, Met1-Gly150, Accession # Q96BF3
0.1 mL
Primary
Human
Purified
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions
Product Content Correction
Product Title
TMIGD2 Antibody (953743), Janelia Fluor™ 646, Novus Biologicals™ > 0.1 mL; Janelia Fluor 646