missing translation for 'onlineSavingsMsg'
Learn More

CH25H Antibody, mFluor Violet 450 SE, Novus Biologicals™

Rabbit Polyclonal Antibody

Brand:  Novus Biologicals NBP3-20218MFV450

Product Code. 18532799

  • 481.00€ / 0.10mL

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

CH25H Polyclonal antibody specifically detects CH25H in Human samples. It is validated for Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY
Specifications

Specifications

CH25H
Polyclonal
Immunohistochemistry-Paraffin
C25H, cholesterol 25-hydroxylase, Cholesterol 25-monooxygenase, EC 1.14.99.38, h25OH
Produced in rabbits immunized with a synthetic peptide corresponding to the C-terminus of the Human CH25H
0.1 mL
Primary
Human
Purified
Immunohistochemistry (Paraffin)
mFluor Violet 450 SE
50mM Sodium Borate
Rabbit
Antigen and protein A Affinity-purified
RUO
9023
Store at 4C in the dark.
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

One moment while we fetch your results.
Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
CH25H Antibody, mFluor Violet 450 SE, Novus Biologicals™ > 0.1 mL; mFluor Violet 450 SE

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.