Learn More
Thermo Scientific™ EPD Mk2, G and N2 Readers
To complement the Thermo Scientific EP DMk2, G and N2. The basic IrDA reader, is ideal for office environments and mobile applications
Brand:  Thermo Scientific™ EPD162570500
 View more versions of this product
Product Code. 11927927
  • 3220.00€ / Each
Explore more special offers
Récupel-bijdrage uitgesloten
Description
Description
Compatible with the full range of Mk2 software products
  • Connect directly to the PC via a serial or optional USB interface
  • Require no power supply
  • For more demanding environments, and where it is necessary to ensure that only one specific EPD can be “seen” by the reader, the desktop model ACT-5 may be used
  • The IrDA and ACT-5 are compatible with EasyEPD2 and EasyIssue software
Specifications
Specifications
0.1 lb.
Serial RS232 or USB 1.1 and 2.0 compatible interface direct to PC
Easy EPD software, USB interface
1 m
50 g
For office Environments and Mobile applications
3.2 ft.
Basic Irda Reader
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Thermo Scientific™ EPD Mk2, G and N2 Readers > Basic IrDA; Easy EPD software; USB interface