Learn More
SI Analytics™ Ubbelohde Viscometers (ASTM)
SI Analytics™ Ubbelohde Viscometers (ASTM) feature a suspended ball level for determination of absolute and relative kinematic viscosity of liquids with Newtonian flow behavior.
Brand:  SI Analytics™ 285400559
Product Code. 10141501
  • 481.00€ / Each
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Calibrated, with constant, for manual measurement
  • Ideal for transparent liquids manual measurement, foaming liquids, liquid mixture with highly volatile components, and high-temperature or low temperature measurements
  • All viscometers are provided with ring marks
  • This ensures that viscometers for automatic measurements can also be checked by means of manual measurements
  • The recommended minimum flowthrough time is 200 seconds
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Ubbelohde Viscometer
Yes
0.1
Glass
Manual measurements
20 to 100 cSt
1.03 mm diamter
ISO 3105, ASTM D 2515, ASTM D 446
285 mm
15 to 20 mL
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Promotions
Promotions
Product Content Correction
Product Title
SI Analytics™ Ubbelohde Viscometers (ASTM) > Range: 20 to 100 mm2/s