Learn More
Thermo Scientific™ Tango™ GPR39-bla U2OS Cells
Tango™ GPR39-bla U2OS cells contain the orphan human G-protein Coupled Receptor 39 (GPR39) linked to a TEV protease site and a Gal4-VP16 transcription factor stably integrated into the Tango™ GPCR-bla U2OS parental cell line.
Brand:  Thermo Scientific™ K1842
Product Code. 10491405
  • 13050.00€ / Each
Estimated Shipment: 02-04-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
Tango™ GPR39-bla U2OS cells contain the orphan human G-protein Coupled Receptor 39 (GPR39) linked to a TEV protease site and a Gal4-VP16 transcription factor stably integrated into the Tango™ GPCR-bla U2OS parental cell line. This parental cell line stably expresses a beta-arrestin/TEV protease fusion protein and the beta-lactamase reporter gene under the control of a UAS response element. The Tango™ GPR39-bla U2OS cells have been functionally validated for a receptor specific response to FBS. In addition, Tango™ GPR39-bla U2OS cells have been tested for assay performance under variable conditions.
TRUSTED_SUSTAINABILITY
Specifications
Show More
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Thermo Scientific™ Tango™ GPR39-bla U2OS Cells > 8 x 106 Cells
For Research Use Only. Not for use in diagnostic procedures.