Learn More
Sampling Systems™ SteriWare™ Disposable Liquid Funnel
Ideal for laboratories, pharmaceutical environments, cleanrooms and chemical manufactruing. Sampling Systems™ SteriWare™ Disposable Liquid Funnel comes individually bagged after being moulded and packed in a Class 100,000 Cleanroom.
Brand: Sampling Systems™ 8370H-100
Description
- Constructed of HDPE
- Moulded and packed in a Class 100,000 Cleanroom
- Individually bagged
- Made from FDA, EC 1935/2004 and EU 10/2011 conforming material
- BSE/TSE free
- Full batch traceability
- Available pre-sterilized (SAL 10-6)
- No cross contamination
- No cleaning required
Specifications
100 mm | |
12 mm | |
Non-sterile | |
Liquid Funnel |
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
108629 | Certificate of Compliance | 15/11/2024 | 8370H-100 |
108392 | Certificate of Compliance | 28/02/2023 | 8370H-100 |
071927 | Certificate of Compliance | 07/12/2022 | 8370H-100 |
060884 | Certificate of Compliance | 16/08/2022 | 8370H-100 |
104419 | Certificate of Compliance | 28/07/2022 | 8370H-100 |