Learn More
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
11937999
![]() |
Mecmesin™
871002 |
1.5 Nm |
POA
Each |
N/A | Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11957999
![]() |
Mecmesin™
871005 |
6 Nm |
POA
Each |
N/A | Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
- A range of fine-precision, low-capacity torque screwdrivers for handheld applications.
- Equipped with 1/4 in. hex drive (or 3mm bore) for the fitting of adaptors.
- Accuracy ±0.5% of the full scale
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Mecmesin™ Static Torque Screwdriver