Learn More
Mecmesin™ Static Torque Screwdriver
Encompass
Suitable for the measurement of very fine static torque.
POA
Products 2
Product Code Brand Capacity (Metric) Price Quantity & Availability  
Product Code Brand Capacity (Metric) Price Quantity & Availability  
11937999
Encompass
View Documents
Mecmesin™
871002
1.5 Nm
POA
Each
View alternative products
Save Up to 
»
N/A
This item requires a price update prior to being ordered.
Request a Price for this item here.
11957999
Encompass
View Documents
Mecmesin™
871005
6 Nm
POA
Each
View alternative products
Save Up to 
»
N/A
This item requires a price update prior to being ordered.
Request a Price for this item here.
Description
Description
  • A range of fine-precision, low-capacity torque screwdrivers for handheld applications.
  • Equipped with 1/4 in. hex drive (or 3mm bore) for the fitting of adaptors.
  • Accuracy ±0.5% of the full scale
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Mecmesin™ Static Torque Screwdriver