Learn More
Mecmesin™ Static Torque Screwdriver
Suitable for the measurement of very fine static torque.
Brand: Mecmesin™ 871005
This item is not returnable.
View return policy
Description
- A range of fine-precision, low-capacity torque screwdrivers for handheld applications.
- Equipped with 1/4 in. hex drive (or 3mm bore) for the fitting of adaptors.
- Accuracy ±0.5% of the full scale
Specifications
6 Nm |