Learn More
Buerkle™ Stainless Steel Safety Container
Buerkle™ Stainless Steel Safety Container offer safety through flame protection and excess pressure valve.
1220.00€ - 2010.00€
Specifications
Material Stainless Steel
View More Specs
Products 4
Product Code Brand Capacity (Metric) Closure Type For Use With (Application) Price Quantity & Availability  
Product Code Brand Capacity (Metric) Closure Type For Use With (Application) Price Quantity & Availability  
15360605
View Documents
Buerkle™
2601-4010
10 L Tap, Pressure Relief Valve Storage and Transfer
1460.00€
Each
Estimated Shipment: 30-04-2025
to see stock.
Add to basket
15941602
View Documents
Buerkle™
2601-6010
10 L Tab, Pressure-relief Valve Storage and Transfer
2010.00€
Each
Estimated Shipment: 29-04-2025
to see stock.
Add to basket
17104165
View Documents
Buerkle™
2601-4025
25 L Tab, Pressure-relief Valve Storage
1730.00€
Each
Estimated Shipment: 27-06-2025
to see stock.
Add to basket
15463375
View Documents
Buerkle™
2601-5050
50 L Screw Cap, Pressure-relief Valve Storage
1220.00€
Each
Estimated Shipment: 27-06-2025
to see stock.
Add to basket
Description
Description
  • Safety containers for storage made of high-quality stainless steel AISI 304 (1.4301) / AISI 316 Ti (1.4571) with thickness of sides of 0.8mm
  • Protection from exploding through flame protection and excess pressure valve, seals made of PTFE, the containers have a high degree of chemical resistance
  • Pressure balance takes place from about 0.3 bar
  • Constant ventilation can be set via a plastic lever on the screw cap
Specifications
Specifications
Stainless Steel
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Product Title
Buerkle™ Stainless Steel Safety Container