Learn More
SP Bel-Art™ Stainless Steel Pharma Scoops
Seamless design for peace of mind
88.95€ - 295.00€
Product Code | Brand | Capacity (English) | Capacity (Metric) | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (English) | Capacity (Metric) | Price | Quantity & Availability | ||||
16814146
|
SP Bel-Art™
F36738-0010 |
0.34 oz. | 10 mL |
88.95€
Each |
Estimated Shipment: 01-08-2024
Log in to see stock. |
||||
16824146
|
SP Bel-Art™
F36738-0050 |
1.7 oz. | 50 mL |
137.00€
Each |
Estimated Shipment: 22-08-2024
Log in to see stock. |
||||
16834146
|
SP Bel-Art™
F36738-0100 |
3.5 oz. | 100 mL |
178.00€
Each |
Estimated Shipment: 22-08-2024
Log in to see stock. |
||||
16844146
|
SP Bel-Art™
F36738-0500 |
17.6 oz. | 500 mL |
192.00€
Each |
Estimated Shipment: 06-09-2024
Log in to see stock. |
||||
16854146
|
SP Bel-Art™
F36738-0750 |
26.4 oz. | 750 mL |
206.00€
Each |
Estimated Shipment: 22-08-2024
Log in to see stock. |
||||
16864146
|
SP Bel-Art™
F36738-1000 |
35.2 oz. | 1000 mL |
233.00€
Each |
Estimated Shipment: 02-08-2024
Log in to see stock. |
||||
16874146
|
SP Bel-Art™
F36738-1250 |
44 oz. | 1250 mL |
240.00€
Each |
Estimated Shipment: 22-08-2024
Log in to see stock. |
||||
16884146
|
SP Bel-Art™
F36738-1700 |
60 oz. | 1700 mL |
254.00€
Each |
Estimated Shipment: 22-08-2024
Log in to see stock. |
||||
16894146
|
SP Bel-Art™
F36738-2500 |
88 oz. | 2500 mL |
295.00€
Each |
Estimated Shipment: 22-08-2024
Log in to see stock. |
||||
Description
- Reusable, high-quality scoops can be used for powder and granule handling in pharmaceutical and food handling settings
- Pharmaceutical grade 316 Stainless Steel
- Easy to clean seamless design to help prevent cross-contamination
- High salt and acid resistance, ideal for food sampling
- GMP compliant
- Autoclavable
Specifications
Stainless Steel |