Learn More
SP Bel-Art™ Stainless Steel Pharma Scoops
Seamless design for peace of mind
88.95€ - 295.00€
Specifications
Material Stainless Steel
View More Specs
Products 9
Product Code Brand Capacity (English) Capacity (Metric) Price Quantity & Availability  
Product Code Brand Capacity (English) Capacity (Metric) Price Quantity & Availability  
16814146
View Documents Promotion Details
SP Bel-Art™
F36738-0010
0.34 oz. 10 mL
88.95€
Each
View alternative products
Save Up to 
»
Estimated Shipment: 01-08-2024
to see stock.
Add to basket
16824146
View Documents Promotion Details
SP Bel-Art™
F36738-0050
1.7 oz. 50 mL
137.00€
Each
View alternative products
Save Up to 
»
Estimated Shipment: 22-08-2024
to see stock.
Add to basket
16834146
View Documents Promotion Details
SP Bel-Art™
F36738-0100
3.5 oz. 100 mL
178.00€
Each
View alternative products
Save Up to 
»
Estimated Shipment: 22-08-2024
to see stock.
Add to basket
16844146
View Documents Promotion Details
SP Bel-Art™
F36738-0500
17.6 oz. 500 mL
192.00€
Each
View alternative products
Save Up to 
»
Estimated Shipment: 06-09-2024
to see stock.
Add to basket
16854146
View Documents Promotion Details
SP Bel-Art™
F36738-0750
26.4 oz. 750 mL
206.00€
Each
View alternative products
Save Up to 
»
Estimated Shipment: 22-08-2024
to see stock.
Add to basket
16864146
View Documents Promotion Details
SP Bel-Art™
F36738-1000
35.2 oz. 1000 mL
233.00€
Each
View alternative products
Save Up to 
»
Estimated Shipment: 02-08-2024
to see stock.
Add to basket
16874146
View Documents Promotion Details
SP Bel-Art™
F36738-1250
44 oz. 1250 mL
240.00€
Each
View alternative products
Save Up to 
»
Estimated Shipment: 22-08-2024
to see stock.
Add to basket
16884146
View Documents Promotion Details
SP Bel-Art™
F36738-1700
60 oz. 1700 mL
254.00€
Each
View alternative products
Save Up to 
»
Estimated Shipment: 22-08-2024
to see stock.
Add to basket
16894146
View Documents Promotion Details
SP Bel-Art™
F36738-2500
88 oz. 2500 mL
295.00€
Each
View alternative products
Save Up to 
»
Estimated Shipment: 22-08-2024
to see stock.
Add to basket
Description
Description
  • Reusable, high-quality scoops can be used for powder and granule handling in pharmaceutical and food handling settings
  • Pharmaceutical grade 316 Stainless Steel
  • Easy to clean seamless design to help prevent cross-contamination
  • High salt and acid resistance, ideal for food sampling
  • GMP compliant
  • Autoclavable