Learn More
SP Bel-Art™ Stainless Steel Pharma Scoops
Seamless design for peace of mind
Brand:  SP Bel-Art™ F36738-1000
Additional Details : Weight : 0.81650kg
 View more versions of this product
Product Code. 16864146
  • 233.00€ / Each
Estimated Shipment: 07-08-2024
to see stock.
Add to basket
Description
Description
  • Reusable, high-quality scoops can be used for powder and granule handling in pharmaceutical and food handling settings
  • Pharmaceutical grade 316 Stainless Steel
  • Easy to clean seamless design to help prevent cross-contamination
  • High salt and acid resistance, ideal for food sampling
  • GMP compliant
  • Autoclavable
Specifications
Specifications
Stainless Steel
35.2 oz.
1000 mL