Learn More
SP Bel-Art™ Stainless Steel Pharma Scoops
Seamless design for peace of mind
Brand: SP Bel-Art™ F36738-1000
Additional Details : Weight : 0.81650kg
Description
- Reusable, high-quality scoops can be used for powder and granule handling in pharmaceutical and food handling settings
- Pharmaceutical grade 316 Stainless Steel
- Easy to clean seamless design to help prevent cross-contamination
- High salt and acid resistance, ideal for food sampling
- GMP compliant
- Autoclavable
Specifications
Stainless Steel | |
35.2 oz. | |
1000 mL |