Learn More
SI Analytics™ Stainless Steel Brackets
Protect your viscometer from breakage. SI Analytics™ Stainless Steel Brackets are designed to ensure that the viscometers are held in a vertical position.
Brand:  SI Analytics™ 285405019
 View more versions of this product
Product Code. 10241881
  • 285.00€ / Each
Estimated Shipment: 16-05-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • The maximum deviation is < 1°
  • In application in conjunction with SCHOTT Instruments and other commercially available see-through thermostats the viscometers can only be used with the appropriate stand or bracket
  • Provide protection for viscometers
  • Allow viscometers to be held vertically
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Bracket
Reverse Flow Viscometer
Stainless Steel
For manual and automatic measurements
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions
Product Content Correction
Product Title
SI Analytics™ Stainless Steel Brackets > For Use With: For All Reverse flow Viscometer