Learn More
Sonoco™ Utek™ Mailing Refrigerant Packs
Phase change materials are ideal for products requiring frozen temperatures or extreme cold that may not tolerate dry ice or CO2
Brand: Sonoco™ 432
Description
- Gel packs have a laminated, metalized film exterior that provides radiant heat barrier, resists punctures
- Single packets are ideal for use as a cubed ice substitute
- Mat is well suited for wraparound applications
Specifications
36 Cs. | |
0.75 in. | |
-23°C | |
6 in. | |
Laminated, Metallized Film | |
227 g | |
10 cm |
U-tek Refrigerant Pack | |
Ideal for products requiring frozen temperatures or extreme cold that may not tolerate dry ice or Carbon dioxide | |
1.9 cm | |
15.2 cm | |
8 oz. | |
4 in. | |
6 x 4 x 0.75 in. |