Learn More
BINDER™ Access Port
Access port with silicone plug for use with KB PRO 130, KB PRO 260, KB PRO 720, KB PRO 1060, and KB PRO 1600.
Brand:  BINDER™ 8012-2475
 View more versions of this product
Product Code. 30238257
  • 943.00€ / Each
Estimated Shipment: 06-05-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Round access port with silicone plug
  • Choose between 30, 50, or 100 mm plug
  • Available on left, right, or top of the chamber
  • For use with KB PRO 130, KB PRO 260, KB PRO 720, KB PRO 1060, and KB PRO 1600
Specifications
Specifications
KB PRO 130, KB PRO 260, KB PRO 720, KB PRO 1060, KB PRO 1600
Access Port
100 mm
Silicone
1 on Left Side
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions
Product Content Correction
Product Title
BINDER™ Access Port > Access Port, 100 mm, Left