Learn More
Deutsch Neumann™ Silicone Rubber Stoppers
For safe and secure closure of laboratory apparatus. Deutsch Neumann™ Silicone Rubber Stoppers are translucent in color.
1.00€ - 82.35€
Specifications
Color Transparent
Material Silicone
View More Specs
Products 8
Product Code Brand Diameter (Metric) Bottom Diameter (Metric) Top Price Quantity & Availability  
Product Code Brand Diameter (Metric) Bottom Diameter (Metric) Top Price Quantity & Availability  
10717781
View Documents
Deutsch Neumann™
1010518
18 mm 24 mm
1.00€
Each
In Stock
to see stock.
Edge
Add to basket
10588321
View Documents
Deutsch Neumann™
1010556
56 mm 65 mm
13.80€
Each
In Stock
to see stock.
Add to basket
10262161
View Documents
Deutsch Neumann™
1010571
71 mm 83 mm
29.60€
Each
Estimated Shipment: 04-03-2025
to see stock.
Add to basket
10373951
View Documents
Deutsch Neumann™
1010564
64.5 mm 75.5 mm
23.10€
Each
Estimated Shipment: 04-03-2025
to see stock.
Add to basket
10046401
View Documents
Deutsch Neumann™
1010579
79 mm 92 mm
40.35€
Each
Estimated Shipment: 06-03-2025
to see stock.
Add to basket
10406321
View Documents
Deutsch Neumann™
1010594
94 mm 107 mm
82.35€
Each
Estimated Shipment: 07-03-2025
to see stock.
Add to basket
10757971
View Documents
Deutsch Neumann™
1010550
50.5 mm 59.5 mm
10.90€
Each
In Stock
to see stock.
Edge
Add to basket
10788351
View Documents
Deutsch Neumann™
1010560
60 mm 70 mm
21.10€
Each
In Stock
to see stock.
Edge
Add to basket
Description
Description
  • Acc. to DIN 12 871
  • Durometer 50 Shore A
  • Available in a wide range of sizes
Specifications
Specifications
Transparent
Silicone
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Product Title
Deutsch Neumann™ Silicone Rubber Stoppers