Learn More
Deutsch Neumann™ Silicone Rubber Stoppers
For safe and secure closure of laboratory apparatus. Deutsch Neumann™ Silicone Rubber Stoppers are translucent in color.
Brand:  Deutsch Neumann™ 1010550
 View more versions of this product
Product Code. 10757971
  • 10.90€ / Each
In Stock
to see stock.
Edge
Explore more special offers
Add to basket
Description
Description
  • Acc. to DIN 12 871
  • Durometer 50 Shore A
  • Available in a wide range of sizes
Specifications
Specifications
1/Ea.
50.5 mm
Silicone
Transparent
59.5 mm
Silicone Stopper