missing translation for 'onlineSavingsMsg'
Learn More

Semi-Volatile Organics Mix A, SPEX CertiPrep™

ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material

Brand:  SPEX Certiprep 5252-A


Product Code. 15245786

  • 125.00€ / 1mL

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers

Includes: Autosampler Vial, NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet
This item is not returnable. View return policy

Description
Show More
Specifications

Specifications

Semi-Volatile Organics Mix A
1 mL
For US EPA Method 525
Acetone
2mL amber prescored glass ampule
Certificate of Analysis reports target analytes to a +/−2% accuracy and correct for raw material purity.
GC, GC/MS, HPLC, LC/MS, and Other Analytical Instrumentation
Product Suggestions

Product Suggestions

Videos
Safety and Handling
Show More
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Semi-Volatile Organics Mix A, SPEX CertiPrep™ > 1mL; 2mL amber prescored glass ampule

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

Product intended for laboratory use only