Learn More
3M™ SecureFit™ SF600 Series Safety Glasses Product Code.: 15787717
Add to basket
3M™ SecureFit™ SF600 Series Safety Glasses
Product Code. 15787717
Scratch-resistant, Polarized, Gray
Click to view available options
Lens Coating:
Anti-fog, Scratch-resistant
Scratch-resistant
Lens Type:
Polarized
Lens Tint:
Amber
Clear
Gray
I/O Mirror
Welding shade 1.7
Welding shade 3.0
Welding shade 5.0
Product Code. 15787717
Brand: 3M™ 7100112713
Edge
Add to basket
Add to basket
Comprised of a semi-rimless lens and fixed-length temple arm. 3M™ SecureFit™ SF600 Series Safety Glasses include an integrated side shield for added protection.
  • Optical class 1 lens suitable for prolonged use
  • Design provides coverage and a good field of vision based on two lenses in a semi-rimless design
  • Offers protection against Ultraviolet (UV) radiation in accordance with European standards
  • Weight: 27g
  • 3M™ Pressure Diffusion Temple Technology for secure and comfortable fit
  • Soft nose pads for comfortable fit
  • Superior Scotchgard™ anti-fog coating meeting K and N on the standard clear, gray, and amber lenses
TRUSTED_SUSTAINABILITY
Specifications
Lens Type Polarized
Lens Coating Scratch-resistant
Lens Tint Gray
Frame Color Gray
Lens Material Polycarbonate
Frame Material Polycarbonate
UV Protection Yes
  • 3M - SecureFit protective eyewear
    Video Player is loading.
    Current Time 0:00
    Duration 2:05
    Loaded: 0%
    Stream Type LIVE
    Remaining Time 2:05
    1x
    • Chapters
    • descriptions off, selected
    • captions off, selected
    • en (Main), selected
    3M SecureFit protective eyewear
  • 3M - SecureFit protective eyewear 600 series
    Protective eyewear that self-adjusts to individual wearer, helping to provide comfort all day.
    Video Player is loading.
    Current Time 0:00
    Duration 0:00
    Loaded: 0%
    Stream Type LIVE
    Remaining Time 0:00
    1x
    • Chapters
    • descriptions off, selected
    • captions off, selected
      3M SecureFit protective eyewear 600 series
    • Multiple Manufacturers
      Save up to 60% on gloves, apparel, and more
      Promo Code: 24131
      Expires2025-06-08
      New Offer
      Shop Now
    Product Content Correction
    Product Title
    3M™ SecureFit™ SF600 Series Safety Glasses >