Description
- Unisex safety glasses offering protection and affordability for site visitors or single use protection
- Design provides maximum clarity, excellent coverage and a wide field of vision
- Contoured flexible temples for a close and comfortable fit
- Anti-scratch coating for increased durability
- Clear lens for excellent protection against Ultraviolet (UV) radiation
- Lens marking: 2C-1.2 3M 1 FT
- Frame marking: 3M EN166 FT CE
Specifications
Specifications
Certifications/Compliance | CE Approved; EN 166:2001 |
Lens Coating | Scratch-resistant |
For Use With (Application) | Offer protection and affordability for site visitors or single-use protection |
Frame Color | Clear |
Temple Style | Flexible |
Lens Material | Polycarbonate |
Product Suggestions
Customers who viewed this item also viewed.
Viewing 1-4 of
Documents
Product Information
Datasheets
7000061918
File size: 226.6 kB
Promotions
-
Multiple Manufacturers
Gear Up for Safety
Save up to 60% on gloves, apparel, and morePromo Code: 24131Expires: 2025-06-08
New Offer Shop Now
Product Content Correction
Product Title
3M™ Virtua™ Safety Glasses, 71500-00001 >