missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SCLY Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
221.00€ - 305.00€
Specifications
Antigen | SCLY |
---|---|
Dilution | Immunohistochemistry-Paraffin 1:50-1:200 |
Applications | Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18665282
|
Novus Biologicals
NBP2-97263-100ul |
100 μg |
305.00€
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18698644
|
Novus Biologicals
NBP2-97263-50ul |
50 μg |
221.00€
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
SCLY Polyclonal antibody specifically detects SCLY in Human samples. It is validated for Immunohistochemistry (Paraffin)Specifications
SCLY | |
Immunohistochemistry (Paraffin) | |
Unconjugated | |
Rabbit | |
Human | |
EC 4.4.1.16, SCLhSCL, selenocysteine lyase | |
Produced in rabbits immunized with E. coli-derived Human SCLY fragment. | |
Primary | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Immunohistochemistry-Paraffin 1:50-1:200 | |
Polyclonal | |
Purified | |
RUO | |
PBS (pH 7.0) | |
51540 | |
IgG | |
Antigen and protein A Affinity-purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
SCLY Antibody, Novus Biologicals™