missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SCLY Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-97263-50ul
This item is not returnable.
View return policy
Description
SCLY Polyclonal antibody specifically detects SCLY in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
SCLY | |
Polyclonal | |
Immunohistochemistry-Paraffin 1:50-1:200 | |
EC 4.4.1.16, SCLhSCL, selenocysteine lyase | |
Produced in rabbits immunized with E. coli-derived Human SCLY fragment. | |
50 μg | |
Primary | |
Human | |
Purified |
Immunohistochemistry (Paraffin) | |
Unconjugated | |
PBS (pH 7.0) | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
51540 | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
IgG |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-4 of
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
SCLY Antibody, Novus Biologicals™ > 50 μg, Unconjugated
Spot an opportunity for improvement?Share a Content Correction