missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Brady™ S3100 Sign and Label Printer- F&B Kit EU
Increase the safety and efficiency of your food or beverage plant with this complete safety sign and label printer kit.
Brand: Brady™ 311116
This item is not returnable.
View return policy
Description
- Easily design and print professional food or beverage safety signs, pipe markers, and labels when you need them
- Print incredibly fast with automatic setup, built-in keyboard, and touchscreen interface
- Create reliable labels that are washdown resistant and do not crumble when exposed to cleaning agents or pressure
- Fully protect food processing with metal detectable signs that enable additional protection against contamination
Specifications
LCD Touchscreen | |
12 in. | |
9.5 in. | |
10 in. | |
Not Autoclavable | |
Monochrome | |
256 MB | |
76 mm/s | |
Brady™ Workstation Basic Design Suite | |
9.1 kg | |
Component and Equipment Labeling, General Identification, Hazard Identification, Temperature Labeling | |
AC Adaptor | |
S3100-QY-EU-FnB |
Thermal Transfer | |
305 mm | |
241 mm | |
260 mm | |
Non-sterile | |
Bulgarian, Chinese (Simplified), Chinese (Tradditional), Croatian, Czech, Danish, Dutch, English, Estonian, Finnish, French, German, Hungarian, Italian, Korean, Norwegian, Polish, Portuguese, Russian, Romanian, Slovakian, Slovenian, Spanish, Swedish, Turkish | |
S3100 | |
300 dpi | |
Ethernet, USB | |
UL, cUL, CE, WEEE, RoHS, RCM, EAC, CCC, RoHS 2 | |
1 Year | |
110/240 V |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-4 of
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Brady™ S3100 Sign and Label Printer- F&B Kit EU > F&B Printer Kit EU; Color Capability: Monochrome; Display: LCD Touchscreen; Depth (Metric): 260 mm; Height (Metric): 241 mm; Width (Metric): 305 mm
Spot an opportunity for improvement?Share a Content Correction