Learn More
Mecmesin™ S-Beam Load Cell
Mecmesin's S-Beam Loadcell is a flexible solution for general force regulation applications.
1045.50€ - 1323.00€
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
11913858
|
Mecmesin™
870009 |
500 N |
1045.50€
Each |
Estimated Shipment: 09-12-2024
Log in to see stock. |
|||||
11968019
|
Mecmesin™
870003 |
10,000 N |
1323.00€
Each |
Estimated Shipment: 09-12-2024
Log in to see stock. |
|||||
Description
- This is suitable for areas where space is not limited.
- The unit features a peak hold of maximum reading at a fast rate of 5000 Hz when used with AFTI display and advanced force gauge.
- The unit is also applicable for use in tension and compression testing.