Learn More
Mecmesin™ S-Beam Load Cell
Mecmesin's S-Beam Loadcell is a flexible solution for general force regulation applications.
1045.50€ - 1323.00€
Products 2
Product Code Brand Capacity (Metric) Price Quantity & Availability  
Product Code Brand Capacity (Metric) Price Quantity & Availability  
11913858
Encompass
View Documents Promotion Details
Mecmesin™
870009
500 N
1045.50€
Each
View alternative products
Save Up to 
»
Estimated Shipment: 09-12-2024
to see stock.
Add to basket
11968019
Encompass
View Documents Promotion Details
Mecmesin™
870003
10,000 N
1323.00€
Each
View alternative products
Save Up to 
»
Estimated Shipment: 09-12-2024
to see stock.
Add to basket
Description
Description
  • This is suitable for areas where space is not limited.
  • The unit features a peak hold of maximum reading at a fast rate of 5000 Hz when used with AFTI display and advanced force gauge.
  • The unit is also applicable for use in tension and compression testing.