Learn More
Mecmesin™ S-Beam Load Cell
Mecmesin's S-Beam Loadcell is a flexible solution for general force regulation applications.
Brand:  Mecmesin™ 870003
Encompass
 View more versions of this product
Product Code. 11968019
  • 1323.00€ / Each
Estimated Shipment: 10-12-2024
to see stock.
Explore more special offers
Add to basket
Récupel-bijdrage uitgesloten
Description
Description
  • This is suitable for areas where space is not limited.
  • The unit features a peak hold of maximum reading at a fast rate of 5000 Hz when used with AFTI display and advanced force gauge.
  • The unit is also applicable for use in tension and compression testing.
Specifications
Specifications
10,000 N
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Special Offers
Special Offers