Learn More
Mecmesin™ S-Beam Load Cell
Mecmesin's S-Beam Loadcell is a flexible solution for general force regulation applications.
Brand: Mecmesin™ 870009
This item is not returnable.
View return policy
Description
- This is suitable for areas where space is not limited.
- The unit features a peak hold of maximum reading at a fast rate of 5000 Hz when used with AFTI display and advanced force gauge.
- The unit is also applicable for use in tension and compression testing.
Specifications
500 N |