Learn More
ROS Antibody (ROS1/8268R), Janelia Fluor™ 549, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-20759JF549
Description
ROS Monoclonal antibody specifically detects ROS in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
ROS | |
Monoclonal | |
Janelia Fluor 549 | |
proto-oncogene tyrosine-protein kinase ROS, c-ros oncogene 1, receptor tyrosine kinase, c-ros-1, MCF3, ROS | |
Recombinant fragment corresponding to cytoplasmic domain of human ROS protein | |
0.1 mL | |
Protein Kinase | |
6098 | |
Store at 4°C in the dark. | |
IgG κ |
Immunohistochemistry (Paraffin) | |
ROS1/8268R | |
50 mM Sodium Borate | |
Rabbit | |
Protein A or G purified | |
RUO | |
Primary | |
Human | |
Purified |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.