missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Recombinant Western Blot Protein Standards (12-90 kDa)
Recombinant Protein
124.00€ - 319.00€
Specifications
Buffer | 125 mM Tris-HCl, pH 6.8, 10 mM DTT, 2 mM EDTA, 2 % (W/V) SDS, 10 % (W/V) Glycerol, 0.01% (W/V) Bromophenol blue |
---|---|
Content And Storage | Store at -20°C. |
For Use With (Application) | Western Blot |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18634970
|
Novus Biologicals™
NBP3-11650-50Tests |
50 Tests |
124.00€
1 set |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18664901
|
Novus Biologicals™
NBP3-11650-250Tests |
250 Tests |
319.00€
1 set |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Specifications
125 mM Tris-HCl, pH 6.8, 10 mM DTT, 2 mM EDTA, 2 % (W/V) SDS, 10 % (W/V) Glycerol, 0.01% (W/V) Bromophenol blue | |
Store at -20°C. | |
Western Blot |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Novus Biologicals™ Recombinant Western Blot Protein Standards (12-90 kDa)