missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human FDXR, mitochondrial Protein

A DNA sequence encoding the Homo sapiens (Human) FDXR, mitochondrial, was expressed in the hosts and tags indicated.

160.00€ - 2485.00€

Specifications

Accession Number NP_001244941.2
Gene ID (Entrez) 2232
Name FDXR, mitochondrial Protein
Regulatory Status Research Use Only
Endotoxin Concentration Not determined.
View More Specs

Products 6
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15909509
View Documents
enQuireBio™
QP6033-ec-10ug
10 μg This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
N/A
15949509
View Documents
enQuireBio™
QP6033-ec-50ug
50 μg This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
N/A
15999499
View Documents
enQuireBio™
QP6033-ec-100ug
100 μg
554.00€
100µg
Please to purchase this item. Need a web account? Register with us today!
15929509
View Documents
enQuireBio™
QP6033-ec-200ug
200 μg This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
N/A
15939509
View Documents
enQuireBio™
QP6033-ec-500ug
500 μg This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
N/A
15919509
View Documents
enQuireBio™
QP6033-ec-1mg
1 mg
2485.00€
1mg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Specifications

NP_001244941.2
FDXR, mitochondrial Protein
Not determined.
Full Length of Isoform 6
Human
Tris-based buffer, 50% glycerol
2232
Research Use Only
FDXR
Recombinant Protein
STQEKTPQIC VVGSGPAGFY TAQHLLKQHP QAHVDIYEKQ PVPFGLVRFG VAPDHPEVKS YGAEDHRALE IPGEELPGVC SARAFVGWYN GLPENQELEP DLSCDTAVIL GQGNVALDVA RILLTPPEHL ERTDITKAAL GVLRQSRVKT VWLVGRRGPL QVAFTIKELR EMIQLPGARP ILDPVDFLGL QDKIKEVPRP RKRLTELLLR TATEKPGPAE AARQASASRA WGLRFFRSPQ QVLPSPDGRR AAGVRLAVTR LEGVDEATRA VPTGDMEDLP CGLVLSSIGY KSRPVDPSVP FDSKLGVIPN VEGRVMDVPG LYCSGWVKRG PTGVIATTMT DSFLTGQMLL QDLKAGLLPS GPRPGYAAIQ ALLSSRGVRP VSFSDWEKLD AEEVARGQGT GKPREKLVDP QEMLRLLGH
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.