missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human FDXR, mitochondrial Protein
A DNA sequence encoding the Homo sapiens (Human) FDXR, mitochondrial, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6033-ec-200ug
This item is not returnable.
View return policy
Specifications
NP_001244941.2 | |
FDXR, mitochondrial Protein | |
Research Use Only | |
FDXR | |
Recombinant Protein | |
E. coli | |
STQEKTPQIC VVGSGPAGFY TAQHLLKQHP QAHVDIYEKQ PVPFGLVRFG VAPDHPEVKS YGAEDHRALE IPGEELPGVC SARAFVGWYN GLPENQELEP DLSCDTAVIL GQGNVALDVA RILLTPPEHL ERTDITKAAL GVLRQSRVKT VWLVGRRGPL QVAFTIKELR EMIQLPGARP ILDPVDFLGL QDKIKEVPRP RKRLTELLLR TATEKPGPAE AARQASASRA WGLRFFRSPQ QVLPSPDGRR AAGVRLAVTR LEGVDEATRA VPTGDMEDLP CGLVLSSIGY KSRPVDPSVP FDSKLGVIPN VEGRVMDVPG LYCSGWVKRG PTGVIATTMT DSFLTGQMLL QDLKAGLLPS GPRPGYAAIQ ALLSSRGVRP VSFSDWEKLD AEEVARGQGT GKPREKLVDP QEMLRLLGH | |
Greater than 80% as determined by SDS-PAGE. |
2232 | |
200 μg | |
Not determined. | |
Full Length of Isoform 6 | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction