missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human FDXR, mitochondrial Protein

A DNA sequence encoding the Homo sapiens (Human) FDXR, mitochondrial, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6033-ec-10ug

 View more versions of this product

Product Code. 15909509

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products »

This item is not returnable. View return policy

Description

Description

Specifications

Specifications

NP_001244941.2
FDXR, mitochondrial Protein
Research Use Only
FDXR
Recombinant Protein
E. coli
STQEKTPQIC VVGSGPAGFY TAQHLLKQHP QAHVDIYEKQ PVPFGLVRFG VAPDHPEVKS YGAEDHRALE IPGEELPGVC SARAFVGWYN GLPENQELEP DLSCDTAVIL GQGNVALDVA RILLTPPEHL ERTDITKAAL GVLRQSRVKT VWLVGRRGPL QVAFTIKELR EMIQLPGARP ILDPVDFLGL QDKIKEVPRP RKRLTELLLR TATEKPGPAE AARQASASRA WGLRFFRSPQ QVLPSPDGRR AAGVRLAVTR LEGVDEATRA VPTGDMEDLP CGLVLSSIGY KSRPVDPSVP FDSKLGVIPN VEGRVMDVPG LYCSGWVKRG PTGVIATTMT DSFLTGQMLL QDLKAGLLPS GPRPGYAAIQ ALLSSRGVRP VSFSDWEKLD AEEVARGQGT GKPREKLVDP QEMLRLLGH
Greater than 80% as determined by SDS-PAGE.
2232
10 μg
Not determined.
Full Length of Isoform 6
Human
His-SUMO
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.