missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RBM27 Antibody, Janelia Fluor™ 646, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB100-60426JF646
This item is not returnable.
View return policy
Description
RBM27 Polyclonal antibody specifically detects RBM27 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
RBM27 | |
Polyclonal | |
Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin | |
RNA-binding protein 27, ARRS1, Psc1, RNA binding motif protein 27 | |
The immunogen recognized by this antibody maps to a region between residue 700 and 750 of human RNA binding motif protein 27 using the numbering given in entry Q9P2N5.2 (GeneID 54439). | |
0.1 mL | |
Primary | |
Human, Mouse | |
Purified |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Janelia Fluor 646 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
54439 | |
Store at 4C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
RBM27 Antibody, Janelia Fluor™ 646, Novus Biologicals™ > 0.1 mL; Janelia Fluor 646
Spot an opportunity for improvement?Share a Content Correction