missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Aurora A/B Antibody (3H1), Janelia Fluor™ 646, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-50043JF646
This item is not returnable.
View return policy
Description
Aurora A/B Monoclonal antibody specifically detects Aurora A/B in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence
Specifications
Aurora A/B | |
Monoclonal | |
Janelia Fluor 646 | |
AIK, AIK2, AIM1, AIM-1, AIRK1, AIRK2, ARK1, ARK2, AURA, AurB, AURKA, AURKB, aurkb-sv1, aurkb-sv2, AYK1, BTAK, IAK1, IPL1, PPP1R47, PPP1R48, STK1, STK12, STK15, STK5, STK6, STK7 | |
Recombinant full length human Aurora A expressed in and purified from E. coli. [UniProt# O14965] | |
0.1 mL | |
Primary | |
Store at 4°C in the dark. | |
IgG1 |
Western Blot, Immunohistochemistry, Immunofluorescence | |
3H1 | |
50 mM Sodium Borate | |
Mouse | |
Protein G purified | |
RUO | |
Human, Mouse, Rat | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Aurora A/B Antibody (3H1), Janelia Fluor™ 646, Novus Biologicals™ > 0.1 mL, Janelia Fluor 646
Spot an opportunity for improvement?Share a Content Correction