Learn More
R&D Systems™ Polystyrene Microplates
high-binding, flat-bottom polystyrene microplates
240.00€
Specifications
Format | Standard |
---|---|
Array | 8 x 12 |
Material | Polystyrene |
No. of Wells | 96 |
Well Shape | Flat Bottom (F-well) |
Description
Specifications
Standard | |
Polystyrene | |
Flat Bottom (F-well) |
8 x 12 | |
96 | |
ELISA |
Product Title
R&D Systems™ Polystyrene Microplates