Learn More
R&D Systems™ Polystyrene Microplates
high-binding, flat-bottom polystyrene microplates
Brand:  R&D Systems™ DY990
 View more versions of this product
Product Code. 30117142
  • 240.00€ / Pack of 25
Explore more special offers
Specifications
Standard
Clear
Polystyrene
96
ELISA
8 x 12
ELISA Microplate
360 μL
Flat Bottom (F-well)
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions
Product Content Correction
Product Title
R&D Systems™ Polystyrene Microplates > Clear