Learn More
Quickfit™ Expansion Socket Adapter
Made of borosilicate glass. Connects large sized ground glass joints with smaller sized ones. With the QuickFit™ expansion socket adaptor.
Brand:  Quickfit XA53
58.21 EUR valid until 2025-04-30
Use promo code "24138" to get your promotional price.
 View more versions of this product
Product Code. 10146454
  • 64.60€ / Pack of 2
Estimated Shipment: 04-08-2025
to see stock.
Explore more special offers
Description
Connect large sized ground glass joints with smaller sized ones with the QuickFit™ expansion socket adaptor
  • Useful for the connection of dissimilar sized ground glass joints
  • Large sized socket at the top and small sized cone at the base
TRUSTED_SUSTAINABILITY
Specifications
Specifications
24/29
Borosilicate Glass
34/35
Useful for the connection of dissimilar sized ground Glass joints
Expansion adapter
Documents
Product Information
Certificates
Promotions
Promotions
  • Multiple Manufacturers
    Use promotion code 24138 to get amazing savings on products for your analysis
    Promo Code: 24138
    Expires2025-04-30
    Days
    6
    Hours
    18
    Minutes
    48
    Seconds
    28
  • Multiple Manufacturers
    Stock up on lab beakers, flasks and more before these deals disappear.
    Promo Code24123
    Expires2025-06-14
    New Offer
    Save Now
Product Content Correction
Product Title
Quickfit™ Expansion Socket Adapter > Cone size: 24/29; Socket size: 34/35