Learn More
Quickfit™ Expansion Socket Adapter
Made of borosilicate glass. Connects large sized ground glass joints with smaller sized ones. With the QuickFit™ expansion socket adaptor.
36.20€ - 324.00€
Specifications
For Use With (Application) | Useful for the connection of dissimilar sized ground Glass joints |
---|---|
Material | Borosilicate Glass |
Product Type | Expansion adapter |
Product Code | Brand | Cone Size | Standard Taper Joint No. | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Cone Size | Standard Taper Joint No. | Price | Quantity & Availability | |||||
12146220
|
Quickfit
XA21 |
14/23 | 19/26 |
40.15€
Pack of 2 |
Estimated Shipment: 06-05-2025
Log in to see stock. |
|||||
10427820
|
Quickfit
XA52 |
19/26 | 34/35 |
63.05€
Pack of 2 |
Estimated Shipment: 16-06-2025
Log in to see stock. |
|||||
12495436
|
Quickfit
XA31 |
14/23 | 24/29 |
38.10€
Pack of 2 |
Estimated Shipment: 16-06-2025
Log in to see stock. |
|||||
12156220
|
Quickfit
XA32 |
19/26 | 24/29 |
48.85€
Pack of 2 |
Estimated Shipment: 07-05-2025
Log in to see stock. |
|||||
11928044
|
Quickfit
XA41 |
14/23 | 29/32 |
58.15€
Pack of 2 |
Estimated Shipment: 07-05-2025
Log in to see stock. |
|||||
12614901
|
Quickfit
XA10 |
10/19 | 14/23 |
55.00€
Pack of 2 |
Estimated Shipment: 16-06-2025
Log in to see stock. |
|||||
11996124
|
Quickfit
XA42 |
19/26 | 29/32 |
59.95€
Pack of 2 |
Estimated Shipment: 29-04-2025
Log in to see stock. |
|||||
10881391
|
Quickfit
XA43 |
24/29 | 29/32 |
82.95€
Pack of 2 |
In Stock
Log in to see stock. ![]() |
|||||
12425446
|
Quickfit
XA54 |
29/32 | 34/35 |
66.20€
Pack of 2 |
Estimated Shipment: 16-06-2025
Log in to see stock. |
|||||
11389093
|
Quickfit
XA64 |
29/32 | 40/38 | N/A | ||||||
10146454
|
Quickfit
XA53 |
24/29 | 34/35 |
64.60€
Pack of 2 |
Estimated Shipment: 08-07-2025
Log in to see stock. |
|||||
15558010
|
Quickfit
XA74 |
29/32 | 45/40 | N/A | ||||||
Description
Connect large sized ground glass joints with smaller sized ones with the QuickFit™ expansion socket adaptor
- Useful for the connection of dissimilar sized ground glass joints
- Large sized socket at the top and small sized cone at the base
Specifications
Useful for the connection of dissimilar sized ground Glass joints | |
Borosilicate Glass | |
Expansion adapter |
Product Title
Quickfit™ Expansion Socket Adapter