Learn More
Applied Biosystems™ QuantStudio 6 Flex package
Facilitates real-time PCR based applications with an upgradable block selection. With a simplified workflow, intuitive software, and touch-screen interface, the QuantStudio™ 6 Flex system offers exceptional reproducibility at a price you can afford.
Marca: Applied Biosystems™ A27164
Descrizione
- Affordable price allows you to obtain a high performance instrument with minimal upfront capital investment
- Compatible with over 8 million TaqMan™ assays for gene expression, genetic variation, gene regulation, or protein expression experiments
- The intelligent design of the QuantStudio™ platform combined with Applied Biosystems™ instrument reliability and accuracy
- Easy-to-use software, responsive touch-screen, and effortless block exchange without the need for any tools
- The QuantStudio™ 6 Flex system accommodates the interchange of a 96-well, 96-well Fast, or 384-well block
- The QuantStudio™ 6 Flex system can also be upgraded to a QuantStudio™ 7 Flex system at a later date, as your needs change

Specifica
PC (Tower) | |
96 | |
54.7 cm | |
1 Year Manufacturer's + 3 Year Field Service repair (with 1 planned maintenance/Year) | |
4°C to 99.9°C | |
96-well: 3.9°C/s | |
Optiflex System Lamp | |
0.2 ml (96-well) | |
38 kg |
0.25°C | |
Touchscreen | |
33.8 cm | |
52.5 cm | |
QuantStudio 6 Flex | |
3.6°C/s (96-well) | |
Excitation 455 to 672 nm, Emission 505 to 723 nm | |
FAM™, SYBR™ Green, VIC™, JOE™, HEX™, TET™, ABY™, NED™, TAMRA™, Cy™3, JUN™, ROX™, Texas Red™, Mustang Purple™, Cy™5, LIZ™, Cy™5.5 | |
96-well |
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.