missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Mouse/Rat Flt-3 Ligand/FLT3L Quantikine ELISA Kit
Mouse/Rat Flt-3 Ligand Quantikine ELISA Kit from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.
Brand: R&D Systems™ MFK00
This item is not returnable.
View return policy
Description
The Mouse/Rat Flt-3 Ligand Quantikine ELISA Kit is a Solid Phase Sandwich ELISA that quantifies mouse, rat Flt-3 Ligand in cell culture supernates (50 ul), serum (50 ul).
Specifications
31.20 - 2000pg/mL | |
HRP | |
Natural and Recombinant Mouse Flt-3 Ligand and natural Rat Flt-3 Ligand | |
14256 | |
Cell Culture Supernates (50uL),Serum (50uL) | |
FL, FLG3L, Flt3 ligand, Flt3L, FLT3LG, fms-related tyrosine kinase 3 ligand, SL cytokine |
5pg/mL | |
Cell Culture Supernatant, Serum | |
ELISA | |
1 x 96-well Plate | |
Store the unopened product at 2° to 8°C. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
R&D Systems™ Mouse/Rat Flt-3 Ligand/FLT3L Quantikine ELISA Kit > One 96-well plate
Spot an opportunity for improvement?Share a Content Correction