missing translation for 'onlineSavingsMsg'
Learn More
Mutexil™ Polypropylene Lab Coats Product Code.: 11765413
In Stock
to see stock.
Add to basket

Mutexil™ Polypropylene Lab Coats

Product Code. 11765413
X-Large, White
Click to view available options
Size:
X-Large
2X-Large
3X-Large
Color:
Blue
White

Product Code. 11765413

Brand: Mutexil™ 4180171XL

In Stock
to see stock.
Edge
Add to basket
Add to basket

Polypropylene lab coats for use in laboratory settings. Available in a variety of sizes.

PP non woven labcoat with press-stud fastening and pockets
  • Disposable
  • PP, 40g/m2
  • For maintenance, production site visits
  • For non hazardous environments

Specifications

Product Type General Purpose Lab Coat
Size X-Large
Gender Unisex
Material Polypropylene
Color White
Cuff Style Elastic
Disposable Single-use
Sterility Sterile
No. of Pockets 2
For Use With (Application) Non-hazardous environments with a high frequency change of clothing
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Mutexil™ Polypropylene Lab Coats >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.