missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mutexil™ Visitor Protective Lab Coats
Click to view available options
Closure Type:
Snaps
Zipper
Size:
X-Large
2X-Large
3X-Large
Description
PP non woven labcoat with press-stud fastening and pockets
- Disposable
- PP, 40g/m2
- For maintenance, production site visits
- For non hazardous environments
Specifications
Specifications
Product Type | Lab Coat |
Size | X-Large |
Material | Polypropylene |
Color | White |
Closure Type | Zipper |
Cuff Style | Elastic |
No. of Pockets | None |
Hazard Rating | For non-hazardous environment |
For Use With (Application) | Agribusiness, industry, communities and restoration |
Packaging Type | 5 bags 0f 10 |
Show More |
Documents
Scientific Resources
Catalogs
MUT002 catalogue mutexil
File size: 6.8 MB
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Mutexil™ Visitor Protective Lab Coats >
Spot an opportunity for improvement?Share a Content Correction