Learn More
Kartell™ Dispolab™ Sample Cup Stoppers
Available in the push-in design.
Brand:  Kartell™ 0251400
Product Code. 12069907
  • 26.00€ / Pack of 1000
Estimated Shipment: 06-05-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
Available in the push-in design.
  • Made from polyethylene
  • For use with Centrifichem™, Technicon™ and Beckman™ analyzers
Specifications
Specifications
Push-on
16mm
Polyethylene
Sample Cup Stopper
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Kartell™ Dispolab™ Sample Cup Stoppers > Diameter: 16mm